MilliporeSigma
  • Chemotaxis of Escherichia coli to norepinephrine (NE) requires conversion of NE to 3,4-dihydroxymandelic acid.
Chemotaxis of Escherichia coli to norepinephrine (NE) requires conversion of NE to 3,4-dihydroxymandelic acid.
Journal of bacteriology (2014-09-04)
Sasikiran Pasupuleti, Nitesh Sule, William B Cohn, Duncan S MacKenzie, Arul Jayaraman, Michael D Manson
ABSTRACT
Norepinephrine (NE), the primary neurotransmitter of the sympathetic nervous system, has been reported to be a chemoattractant for enterohemorrhagic Escherichia coli (EHEC). Here we show that nonpathogenic E. coli K-12 grown in the presence of 2 μM NE is also attracted to NE. Growth with NE induces transcription of genes encoding the tyramine oxidase, TynA, and the aromatic aldehyde dehydrogenase, FeaB, whose respective activities can, in principle, convert NE to 3,4-dihydroxymandelic acid (DHMA). Our results indicate that the apparent attractant response to NE is in fact chemotaxis to DHMA, which was found to be a strong attractant for E. coli. Only strains of E. coli K-12 that produce TynA and FeaB exhibited an attractant response to NE. We demonstrate that DHMA is sensed by the serine chemoreceptor Tsr and that the chemotaxis response requires an intact serine-binding site. The threshold concentration for detection is ≤5 nM DHMA, and the response is inhibited at DHMA concentrations above 50 μM. Cells producing a heterodimeric Tsr receptor containing only one functional serine-binding site still respond like the wild type to low concentrations of DHMA, but their response persists at higher concentrations. We propose that chemotaxis to DHMA generated from NE by bacteria that have already colonized the intestinal epithelium may recruit E. coli and other enteric bacteria that possess a Tsr-like receptor to preferred sites of infection.
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%