MilliporeSigma
Detection of the alpha-subunit of inhibin in trophoblastic neoplasia.
Human pathology (1999-01-29)
T J Pelkey, H F Frierson, S E Mills, M H Stoler
ABSTRACT
Placental trophoblasts are the primary source of serum inhibin during pregnancy. We sought to characterize inhibin immunolabeling in trophoblastic neoplasms and compare the results with those for established markers of trophoblastic differentiation. Formalin-fixed, paraffin-embedded tissues from three normal term placentas, 13 hydatidiform moles (HM), five choriocarcinomas (CC) (three gestational, one testicular, one mediastinal), and five placental site trophoblastic tumors (PSTT) were immunolabeled with antibodies to the alpha-subunit of inhibin, hPL, and beta-hCG. Additionally, six first-trimester placentas were immunolabeled with antibody to inhibin alone. Trophoblastic subpopulations were assessed for the number of positive cells (1% to 24% = 1+,25% to 49% = 2+,50% to 74% = 3+, 75% to 100% = 4+). Immunolabeling in term placenta and HM was similar, because beta-hCG was the most sensitive marker (4+) of syncytiotrophoblasts followed by inhibin (3-4+) and hPL (2-3+). Immunolabeling of syncytiotrophoblasts in CC was similar to that in term placenta and HM, except for negative hPL staining in two cases. In HM, inhibin labeling of intermediate trophoblasts (2-3+) was less than that for hPL (3-4+). In CC, inhibin and hPL labeling of intermediate trophoblasts was more variable with negative hPL reactivity in two cases. Inhibin and hPL did not stain cytotrophoblasts. In PSTT, inhibin immunolabeling was a better marker than hPL in two cases, inferior in two, and similar in one. Inhibin labeling of syncytiotrophoblasts was less than that for beta-hCG, but did not have the often marked background staining that was present with beta-hCG. Immunolabeling of trophoblast subpopulations for inhibin was similar between first-trimester placenta, term placenta, and HM. We conclude that inhibin is a useful immunohistochemical marker of trophoblastic neoplasia and should be included in the antibody panel with beta-hCG and hPL.
Sigma-Aldrich
Inhibin, alpha (R1) Mouse Monoclonal Antibody