MilliporeSigma
  • Myeloid sarcoma: clinico-pathologic, phenotypic and cytogenetic analysis of 92 adult patients.
Myeloid sarcoma: clinico-pathologic, phenotypic and cytogenetic analysis of 92 adult patients.
Leukemia (2006-12-16)
S A Pileri, S Ascani, M C Cox, C Campidelli, F Bacci, M Piccioli, P P Piccaluga, C Agostinelli, S Asioli, D Novero, M Bisceglia, M Ponzoni, A Gentile, P Rinaldi, V Franco, D Vincelli, A Pileri, R Gasbarra, B Falini, P L Zinzani, M Baccarani
ABSTRACT
Myeloid sarcoma (MS) is a rare neoplasm whose knowledge is largely based on case reports and/or technically dated contributions. Ninety-two MSs in adulthood with clinical data available were evaluated both morphologically and immunohistochemically. Seventy-four cases were also studied by fluorescent in situ hybridization on tissue sections and/or conventional karyotyping on bone marrow or peripheral blood. Histologically, 50% of the tumors were of the blastic type, 43.5% either monoblastic or myelomonocytic and 6.5% corresponded to different histotypes. CD68/KP1 was the most commonly expressed marker (100%), followed by myeloperoxidase (83.6%), CD117 (80.4%), CD99 (54.3%), CD68/PG-M1 (51%), CD34 (43.4%), terminal-deoxy-nucleotidyl-transferase (31.5%), CD56 (13%), CD61/linker for activation of T cells (2.2%), CD30 (2.2%) and CD4 (1.1%). Foci of plasmacytoid monocyte differentiation were observed in intestinal cases carrying inv16. Chromosomal aberrations were detected in about 54% of cases: monosomy 7(10.8%), trisomy 8(10.4%) and mixed lineage leukemia-splitting (8.5%) were the commonest abnormalities, whereas t(8;21) was rare (2.2%). The behavior was dramatic irrespective of presentation, age, sex, phenotype and cytogenetics. Most if not all, long survivors received bone-marrow transplantation. The present report expands the spectrum of our knowledge showing that MS has frequent monoblastic/myelomonocytic differentiation, displays distinctive phenotypic profile, carries chromosomal aberrations other than t(8;21), and requires supra-maximal therapy.
Sigma-Aldrich
CD13 (EP117) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
CD13 (SP187) Rabbit Monoclonal Antibody