MilliporeSigma
p21 as a prognostic factor in non-small cell lung carcinomas.
Pathology, research and practice (2006-11-04)
Mi Seon Kwon, Young Il Lee, Kyo Young Lee
ABSTRACT
Few studies have focused on the correlation between p21 expression and survival for patients with non-small cell lung carcinoma (NSCLC), and the results are not consistent. We investigated the expression of p21 in 90 cases of NSCLC to evaluate the correlation between the p21 expression level and the clinicopathologic characteristics with patient survival. p21 was expressed in the nuclei of all the NSCLCs. The percentage of immunoreactive cells varied from 1% to 70%. All the patients were subdivided into a high and a low p21 expression group on the basis of the median percentage (17.05). There was no significant correlation between the p21 expression level and age, gender, histologic type, histologic grade, or stage. Using uni- and multivariate analyses, survival was significantly associated with gender, stage, and the p21 expression level. The survival rate for the high p21 expression group was higher than that for the low p21 expression group in the entire patient group, and especially for stage II and III patients, males, adenocarcinomas, or p53-positive tumors. Our findings showed that high p21 expression was an independent prognostic factor for NSCLC. p21 may be useful for determining the candidates for adjuvant therapies and also for selecting the appropriate chemotherapeutic agents.
Sigma-Aldrich
p21WAF1 (DCS-60.2) Mouse Monoclonal Antibody