MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pulmonary adenocarcinomas with enteric differentiation: histologic and immunohistochemical characteristics compared with metastatic colorectal cancers and usual pulmonary adenocarcinomas.
Pulmonary adenocarcinomas with enteric differentiation: histologic and immunohistochemical characteristics compared with metastatic colorectal cancers and usual pulmonary adenocarcinomas.
The American journal of surgical pathology (2005-04-16)
Kentaro Inamura, Yukitoshi Satoh, Sakae Okumura, Ken Nakagawa, Eiju Tsuchiya, Masashi Fukayama, Yuichi Ishikawa
ABSTRACT
Primary pulmonary adenocarcinomas with enteric differentiation (PAED) are mainly composed of tall-columnar cells that show similarity to intestinal epithelia and colorectal carcinomas. In this study, we analyzed the immunostaining profiles of 7 PAEDs in comparison with 14 metastatic colorectal carcinomas (MCRs) and 30 usual pulmonary adenocarcinomas (PACs), using antibodies against CDX-2, cytokeratin 7 (CK7), cytokeratin 20 (CK20), TTF-1, surfactant apoprotein-A (SP-A), Napsin A, and MUC2. The positive rates for CDX-2, CK7, CK20, TTF-1, SP-A, Napsin A, and MUC2 were 71%, 100%, 43%, 43%, 14%, 0%, and 43%, respectively, in the PAEDs; 100%, 0%, 86%, 0%, 0%, 0%, and 57% in the MCRs; 3%, 100%, 0%, 93%, 73%, 90%, and 0% in PACs. As expected, immunoreactivity of CDX-2, CK20, and MUC2 was detected in PAEDs. The observed decrease or loss of immunoreactivity for TTF-1, SP-A, and Napsin A indicates that these lesions demonstrate a shift away from their pulmonary phenotype, although CK7 expression was retained. The results indicate that CK7 and CK20 may be useful markers for distinction of PAEDs from MCRs.
Sigma-Aldrich
Napsin A (EP205) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
Napsin A Rabbit Polyclonal Antibody
Sigma-Aldrich
Napsin A (MRQ-60) Mouse Monoclonal Antibody