MilliporeSigma
  • Heffalumps, jagulars, and cheshire cats. A commentary on cytokeratins and soft tissue sarcomas.
Heffalumps, jagulars, and cheshire cats. A commentary on cytokeratins and soft tissue sarcomas.
American journal of clinical pathology (1991-04-01)
P E Swanson
ABSTRACT
Historically, antibodies to cytokeratin intermediate filaments have been models of target specificity. In most diagnostic settings, the utility of these antibodies was unquestioned; reactivity for cytokeratin was dogmatically equated with epithelial differentiation. Recently, however, the diagnostic importance of these antibodies has been challenged, prompted by the demonstration of cytokeratin reactivity in a variety of "nonepithelial" neoplasms. In this review, the evolving literature on this topic is explored, and the practical implications of these findings are discussed.
Sigma-Aldrich
Cytokeratin, HMW (AE3) Mouse Monoclonal Antibody
Sigma-Aldrich
Cytokeratin, LMW (AE1) Mouse Monoclonal Antibody