MilliporeSigma
  • p120, a novel substrate of protein tyrosine kinase receptors and of p60v-src, is related to cadherin-binding factors beta-catenin, plakoglobin and armadillo.
p120, a novel substrate of protein tyrosine kinase receptors and of p60v-src, is related to cadherin-binding factors beta-catenin, plakoglobin and armadillo.
Oncogene (1992-12-01)
A B Reynolds, L Herbert, J L Cleveland, S T Berg, J R Gaut
ABSTRACT
A novel protein tyrosine kinase (PTK) substrate, p120, has been previously implicated in ligand-induced signaling through the epidermal growth factor, platelet-derived growth factor and colony-stimulating factor 1 receptors, and in cell transformation by p60v-src. We have isolated a near full-length cDNA encoding murine p120. The encoded protein lacks significant homology with any reported protein, but it contains four copies of an imperfect 42 amino acid repeat that occurs 12.5 times in the protein encoded by Drosophila armadillo (arm), and its direct homologs, human plakoglobin (plak) and Xenopus laevis beta-catenin (beta-cat). The presence of this motif implies that p120 may share at least one aspect of its function with the arm protein and its homologs.
Sigma-Aldrich
p120 Catenin (MRQ-5) Mouse Monoclonal Antibody