MilliporeSigma
An overview of the serpin superfamily.
Genome biology (2006-06-02)
Ruby H P Law, Qingwei Zhang, Sheena McGowan, Ashley M Buckle, Gary A Silverman, Wilson Wong, Carlos J Rosado, Chris G Langendorf, Rob N Pike, Philip I Bird, James C Whisstock
ABSTRACT
Serpins are a broadly distributed family of protease inhibitors that use a conformational change to inhibit target enzymes. They are central in controlling many important proteolytic cascades, including the mammalian coagulation pathways. Serpins are conformationally labile and many of the disease-linked mutations of serpins result in misfolding or in pathogenic, inactive polymers.
Sigma-Aldrich
C1 Esterase inhibitor from human plasma, ≥95% (SDS-PAGE)
Sigma-Aldrich
C1 Inhibitor human, recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
Antithrombin III from human plasma, ≥95% (SDS-PAGE)