MilliporeSigma
  • Development and clinical application of high performance liquid chromatography for the simultaneous determination of plasma levels of theophylline and its metabolites without interference from caffeine.
Development and clinical application of high performance liquid chromatography for the simultaneous determination of plasma levels of theophylline and its metabolites without interference from caffeine.
Biomedical chromatography : BMC (1999-04-07)
J Kizu, S Watanabe, N Yasuno, Y Arakawa, S Uzu, S Kanda, F Komoda, T Iwata, H Hayakawa, T Hayakawa, K Imai
ABSTRACT
A high performance liquid chromatography (HPLC) method has been developed for the simultaneous determination of plasma levels of theophylline and its metabolites without interference from caffeine or caffeine metabolites. The method is simple and of practical use because it is applicable even to plasma samples from patients who take caffeine-containing beverages. The method was also reproducible with a coefficient of variation of less than 5% for each analyte. The levels of theophylline, determined by HPLC, were validated by their high correlation to the levels obtained by fluorescence polarization immunoassay. HPLC was used to determine theophylline levels in patients with bronchial asthma. The data revealed that the ratio of 1,3-dimethyluric acid, the major metabolite of theophylline, to theophylline concentration in the plasma was within a narrow range in most patients (0.055 +/- 0.01, n = 66), regardless of the method of theophylline administration or the time of blood sampling. Conversely, this ratio was as low as 0.027 +/- 0.005 in the patient with a long plasma half-life of theophylline. These results suggest that it may be possible to predict the plasma half-life of theophylline for each patient from a single blood sample. This may be useful when planning theophylline administration, especially in patients with abnormal theophylline metabolism.
Sigma-Aldrich
1,7-Dimethyluric acid, ≥97.0% (HPLC)