MilliporeSigma
  • HGAL, a germinal center specific protein, decreases lymphoma cell motility by modulation of the RhoA signaling pathway.
HGAL, a germinal center specific protein, decreases lymphoma cell motility by modulation of the RhoA signaling pathway.
Blood (2010-09-17)
Xiaoyu Jiang, Xiaoqing Lu, George McNamara, Xiaofei Liu, Elena Cubedo, Kristopher A Sarosiek, Isidro Sánchez-García, David M Helfman, Izidore S Lossos
ABSTRACT
HGAL is a germinal center (GC)-specific gene that negatively regulates lymphocyte motility and whose expression predicts improved survival of patients with diffuse large B-cell lymphoma (DLBCL) and classical Hodgkin lymphoma (cHL). We demonstrate that HGAL serves as a regulator of the RhoA signaling pathway. HGAL enhances activation of RhoA and its down-stream effectors by a novel mechanism - direct binding to the catalytic DH-domain of the RhoA-specific guanine nucleotide exchange factors (RhoGEFs) PDZ-RhoGEF and LARG that stimulate the GDP-GTP exchange rate of RhoA. We delineate the structural domain of HGAL that mediates its interaction with the PDZ-RhoGEF protein. These observations reveal a novel molecular mechanism underlying the inhibitory effects of GC-specific HGAL protein on the motility of GC-derived lymphoma cells. This mechanism may underlie the limited dissemination and better outcome of patients with HGAL-expressing DLBCL and cHL.