MilliporeSigma
  • Dimeric dUTPases, HisE, and MazG belong to a new superfamily of all-alpha NTP pyrophosphohydrolases with potential "house-cleaning" functions.
Dimeric dUTPases, HisE, and MazG belong to a new superfamily of all-alpha NTP pyrophosphohydrolases with potential "house-cleaning" functions.
Journal of molecular biology (2005-03-03)
Olga V Moroz, Alexey G Murzin, Kira S Makarova, Eugene V Koonin, Keith S Wilson, Michael Y Galperin
ABSTRACT
Structure-guided analysis of the new dimeric dUTPase family revealed its sequence relationship to the phage T4 dCTPase, phosphoribosyl-ATP pyrophosphatase HisE, NTP pyrophosphatase MazG, and several uncharacterized protein families, including the human protein XTP3TPA (RS21-C6), which is overexpressed in embryonic and cancer cells. Comparison with the recently determined structure of a MazG-like protein from Sulfolobus solfataricus supported the unification of these enzymes in one superfamily of all-alpha NTP pyrophosphatases, suggesting that dimeric dUTPases evolved from a tetrameric MazG-like ancestor by gene duplication. Analysis of the structure of the Sulfolobus MazG points to 2-hydroxyadenosine (isoguanosine) triphosphate, a product of oxidative damage of ATP, as the most likely substrate. We predict that uncharacterized members of this superfamily perform "house-cleaning" functions by hydrolyzing abnormal NTPs and are functionally analogous to the structurally unrelated hydrolases of the Nudix superfamily. We outline probable tertiary and quaternary structures of the all-alpha NTP pyrophosphatase superfamily members.