MilliporeSigma
  • Cooperative effects of Th2 cytokines and allergen on normal and asthmatic bronchial epithelial cells.
Cooperative effects of Th2 cytokines and allergen on normal and asthmatic bronchial epithelial cells.
Journal of immunology (Baltimore, Md. : 1950) (2002-06-22)
James L Lordan, Fabio Bucchieri, Audrey Richter, Athanassias Konstantinidis, John W Holloway, Matthew Thornber, Sarah M Puddicombe, Diana Buchanan, Susan J Wilson, Ratko Djukanović, Stephen T Holgate, Donna E Davies
ABSTRACT
In sensitized individuals, exposure to allergens such as Dermatophagoides pteronyssinus (Der p) causes Th2 polarization and release of cytokines, including IL-4 and IL-13. Because Der p extracts also have direct effects on epithelial cells, we hypothesized that allergen augments the effects of Th2 cytokines by promoting mediator release from the bronchial epithelium in allergic asthma. To test our hypothesis, primary bronchial epithelial cultures were grown from bronchial brushings of normal and atopic asthmatic subjects. RT-PCR showed that each culture expressed IL-4R(alpha), common gamma-chain, and IL-13R(alpha)(1), as well as IL-13R(alpha)(2), which negatively regulates IL-13 signaling; FACS analysis confirmed IL-13R(alpha)(2) protein expression. Exposure of epithelial cultures to either Der p extracts, TNF-alpha, IL-4, or IL-13 enhanced GM-CSF and IL-8 release, and this was partially suppressible by corticosteroids. Simultaneous exposure of the epithelial cultures to IL-4 or IL-13 together with Der p resulted in a further increase in cytokine release, which was at least additive. Release of TGF-alpha was also increased by TNF-alpha and combinations of IL-4, IL-13, and Der p; however, this stimulation was only significant in the asthma-derived cultures. These data suggest that, in an allergic environment, Th2 cytokines and allergen have the potential to sustain airway inflammation through a cooperative effect on cytokine release by the bronchial epithelium. Our novel finding that IL-4, IL-13, and allergen enhance release of TGF-alpha, a ligand for the epidermal growth factor receptor that stimulates fibroblast proliferation and goblet cell differentiation, provides a potential link between allergen exposure, Th2 cytokines, and airway remodelling in asthma.
Sigma-Aldrich
JumpStart Taq DNA Polymerase, without MgCl2