MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PIAS1 interacts with the KRAB zinc finger protein, ZNF133, via zinc finger motifs and regulates its transcriptional activity.
PIAS1 interacts with the KRAB zinc finger protein, ZNF133, via zinc finger motifs and regulates its transcriptional activity.
Experimental & molecular medicine (2007-10-16)
Sang-Jin Lee, Jae-Rin Lee, Hwa-Sun Hahn, Young-Hoon Kim, Jin-Hyun Ahn, Chang-Dae Bae, Jun-Mo Yang, Myong-Joon Hahn
ABSTRACT
Zinc finger protein 133 (ZNF133) is composed of a Krüppel-associated box (KRAB) domain and 14 contiguous zinc finger motifs. ZNF133 is regarded as a transcriptional repressor because the KRAB domain has potent repressor activity and the zinc finger motifs usually act in binding to DNA. However, we found that the zinc finger motifs of ZNF133 also possessed transcriptional repressor activity. By two-hybrid screening assay, we found that the zinc finger motifs of ZNF133 interacted with protein inhibitor of activated STAT1 (PIAS1). PIAS1 enhanced the transcriptional repression activity of ZNF133 through the zinc finger motifs. This effect of PIAS1 was relieved by an inhibitor of the histone deacetylases (HDACs). These results demonstrate that the transcriptional repressor activity of ZNF133 is regulated by both the KRAB domain and the zinc finger motifs, and that the repressive effect by zinc finger motifs is mediated by PIAS1.