MilliporeSigma
  • Expression of cancer/testis antigens is correlated with improved survival in glioblastoma.
Expression of cancer/testis antigens is correlated with improved survival in glioblastoma.
Oncotarget (2013-04-18)
Marcelo Freitas, Suzana Malheiros, João Norberto Stávale, Thais Priscila Biassi, Fernando Tadeu Zamunér, Maria de Souza Begnami, Fernando Augusto Soares, Andre Luíz Vettore
ABSTRACT
Glioblastoma (GBM) confers a dismal prognosis despite advances in current therapy. Cancer-testis antigens (CTA) comprise families of tumor-associated antigens that are immunogenic in different cancers. The aim of this study was to determine the expression profile of a large number of CTA genes in GBM. We selected, from 153 CTA genes, those genes potentially expressed in GBM. The expression pattern of 30 CTA was then evaluated by RT-PCR in a series of 48 GBM and 5 normal brain samples. The presence of CTCFL protein was also evaluated by immunohistochemical staining. Among the genes with no expression in normal brain, ACTL8 (57%), OIP5 (54%), XAGE3 (44%) and CTCFL (15%) were frequently expressed in GBM, while over 85% of the tumors expressed at least 1 of these four CTA. Coexpression of two or more CTA occurred in 49% of cases. CTCFL protein expression was detected in 13% of the GBM and was negative in normal brain samples. GBM expressing 3-4 CTA was associated with significantly better overall survival (OS) rates (P = 0.017). By multivariate analysis, mRNA positivity for 3-4 CTA (P = 0.044), radiotherapy (P = 0.010) and chemotherapy (P = 0.001) were independent prognostic factors for OS. GBM frequently express ACTL8, OIP5, XAGE3 and CTCFL. A relatively high percentage of tumors expressed at least one of these four CTA, opening the perspective for their utility in antigen-specific immunotherapy. Furthermore, mRNA positivity for 3-4 CTA is an independent predictor of better OS for GBM patients.