MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Anaerobic ammonium oxidation measured in sediments along the Thames estuary, United Kingdom.
Anaerobic ammonium oxidation measured in sediments along the Thames estuary, United Kingdom.
Applied and environmental microbiology (2003-11-07)
Mark Trimmer, Joanna C Nicholls, Bruno Deflandre
ABSTRACT
Until recently, denitrification was thought to be the only significant pathway for N(2) formation and, in turn, the removal of nitrogen in aquatic sediments. The discovery of anaerobic ammonium oxidation in the laboratory suggested that alternative metabolisms might be present in the environment. By using a combination of (15)N-labeled NH(4)(+), NO(3)(-), and NO(2)(-) (and (14)N analogues), production of (29)N(2) and (30)N(2) was measured in anaerobic sediment slurries from six sites along the Thames estuary. The production of (29)N(2) in the presence of (15)NH(4)(+) and either (14)NO(3)(-) or (14)NO(2)(-) confirmed the presence of anaerobic ammonium oxidation, with the stoichiometry of the reaction indicating that the oxidation was coupled to the reduction of NO(2)(-). Anaerobic ammonium oxidation proceeded at equal rates via either the direct reduction of NO(2)(-) or indirect reduction, following the initial reduction of NO(3)(-). Whether NO(2)(-) was directly present at 800 micro M or it accumulated at 3 to 20 micro M (from the reduction of NO(3)(-)), the rate of (29)N(2) formation was not affected, which suggested that anaerobic ammonium oxidation was saturated at low concentrations of NO(2)(-). We observed a shift in the significance of anaerobic ammonium oxidation to N(2) formation relative to denitrification, from 8% near the head of the estuary to less than 1% at the coast. The relative importance of anaerobic ammonium oxidation was positively correlated (P < 0.05) with sediment organic content. This report of anaerobic ammonium oxidation in organically enriched estuarine sediments, though in contrast to a recent report on continental shelf sediments, confirms the presence of this novel metabolism in another aquatic sediment system.
Product Description
SGE Gas Tight Syringes, Luer Lock, 50MR-LL-GT, volume 50 mL, needle size (not included)
SGE Gas Tight Syringes, Luer Lock, 100MR-LL-GT, volume 100 mL, needle size (not included)