MilliporeSigma
  • Reaction network governing diphosphine-protected gold nanocluster formation from nascent cationic platforms.
Reaction network governing diphosphine-protected gold nanocluster formation from nascent cationic platforms.
Physical chemistry chemical physics : PCCP (2012-02-18)
John M Pettibone, Jeffrey W Hudgens
ABSTRACT
We identify the reaction network governing gold monolayer protected cluster (MPC) formation during the reduction of Au(PPh(3))Cl and L(5) (L(5) = 1,5-bis(diphenylphosphino)pentane) in solutions. UV-vis spectroscopy and electrospray ionization mass spectrometry (ESI-MS) monitored the formation of ligated Au(x): 6 ≤ x ≤ 12 clusters, which comprise the reaction intermediates and final products. Initially, predominantly [Au(2)L(5)(2)](2+) complexes form through dissolution of Au(PPh(3))Cl. These complexes control the reduction and nucleation reactions that form nascent phosphine-ligated Au(8) and Au(10) ionic clusters. [Au(10)L(5)(4)](2+) is an observed growth platform for ligated Au(11) and Au(12) clusters. The data for syntheses of Au : L(5) systems evidence that the nascent reaction products (t < 3 days) are less dependent on the chosen reducing agent (borane tert-butylamine complex or NaBH(4)); instead, after reduction ceases, subsequent solution phase processing provides greater control for tuning cluster nuclearity.
Sigma-Aldrich
Borane tert-butylamine complex, powder, 97%