MilliporeSigma
  • Secondary structure and dosage of soluble and membrane proteins by attenuated total reflection Fourier-transform infrared spectroscopy on hydrated films.
Secondary structure and dosage of soluble and membrane proteins by attenuated total reflection Fourier-transform infrared spectroscopy on hydrated films.
European journal of biochemistry (1990-10-24)
E Goormaghtigh, V Cabiaux, J M Ruysschaert
ABSTRACT
Attenuated total reflection Fourier-transform infrared spectroscopy of thin hydrated films of soluble and membrane protein included in a phospholipid bilayer is shown to provide useful information as to the secondary structure of the protein. The analysis of the amide I band of deuterated samples by Fourier self-deconvolution followed by a curve fitting was performed by a new procedure in which all the input parameters are generated by the computer rather than by the investigator. The results of this analysis provide a correct estimation of the alpha-helix and beta-sheet structure content with a standard deviation of 8.6% when X-ray structures are taken as a reference. We also show that the orientation of the different secondary structures resolved by the Fourier self-deconvolution/curve-fitting procedure and of the phospholipid acyl chains can be simultaneously evaluated for membrane proteins reconstituted in a lipid bilayer. Of special interest for reconstitution of membrane proteins, the lipid/protein ratio can be accurately and quickly determined from the infrared spectrum.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
α-Chymotrypsinogen A from bovine pancreas, essentially salt-free, lyophilized powder
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease S from bovine pancreas, Grade XII-S
Sigma-Aldrich
Elastase from porcine pancreas, Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein
Sigma-Aldrich
Trypsinogen from bovine pancreas, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein (E1%/280, after activation to trypsin)
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein