MilliporeSigma
  • L-fucose utilization provides Campylobacter jejuni with a competitive advantage.
L-fucose utilization provides Campylobacter jejuni with a competitive advantage.
Proceedings of the National Academy of Sciences of the United States of America (2011-04-13)
Martin Stahl, Lorna M Friis, Harald Nothaft, Xin Liu, Jianjun Li, Christine M Szymanski, Alain Stintzi
ABSTRACT
Campylobacter jejuni is a prevalent gastrointestinal pathogen in humans and a common commensal of poultry. When colonizing its hosts, C. jejuni comes into contact with intestinal carbohydrates, including L-fucose, released from mucin glycoproteins. Several strains of C. jejuni possess a genomic island (cj0480c-cj0490) that is up-regulated in the presence of both L-fucose and mucin and allows for the utilization of L-fucose as a substrate for growth. Strains possessing this genomic island show increased growth in the presence of L-fucose and mutation of cj0481, cj0486, and cj0487 results in the loss of the ability to grow on this substrate. Furthermore, mutants in the putative fucose permease (cj0486) are deficient in fucose uptake and demonstrate a competitive disadvantage when colonizing the piglet model of human disease, which is not paralleled in the colonization of poultry. This identifies a previously unrecorded metabolic pathway in select strains of C. jejuni associated with a virulent lifestyle.