MilliporeSigma
BMI-1026 treatment can induce SAHF formation by activation of Erk1/2.
BMB reports (2008-08-07)
Hyun-Joo Seo, Hye Jeong Park, Hyung Su Choi, So-Yoon Hwang, Jeong-Soo Park, Yeon-Sun Seong
ABSTRACT
BMI-1026 is a synthetic aminopyrimidine compound that targets cyclin dependent kinases (cdks) and was initially designed as a potential anticancer drug. Even though it has been well documented that BMI-1026 is a potent cdk inhibitor, little is known about the cellular effects of this compound. In this study, we examined the effects of BMI-1026 treatment on inducing premature senescence and then evaluated the biochemical features of BMI-1026-induced premature senescence. From these experiments we determined that BMI-1026 treatment produced several biochemical features of premature senescence and also stimulated expression of mitogen activated protein kinase (MAPK) family proteins. BMI-1026 treatment caused nuclear translocation of activated Erk1/2 and the formation of senescence associated heterochromatin foci in 5 days. The heterochromatin foci formation was perturbed by inhibition of Erk1/2 activation.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000