MilliporeSigma
  • Quantitative analysis of plasma acylcarnitines using gas chromatography chemical ionization mass fragmentography.
Quantitative analysis of plasma acylcarnitines using gas chromatography chemical ionization mass fragmentography.
Journal of lipid research (1997-01-01)
C G Costa, E A Struys, A Bootsma, H J ten Brink, L Dorland, I Tavares de Almeida, M Duran, C Jakobs
ABSTRACT
A stable isotope dilution gas chromatography chemical ionization mass spectrometry (GC-CI-MS) method was developed for the quantitative profiling of plasma acylcarnitines. The clean-up procedure was comprised of a solid-phase cation exchange extraction using PRS-columns from which the acylcarnitines were eluted with a barium chloride solution. Isolated acylcarnitines were transformed into acyloxylactones and analyzed by positive GC-CI-MS using isobutane as reactant gas. The selected monitoring of a common ion at m/z [85]+ and the protonated molecular ion enabled a selective and sensitive detection of all C2-C18 acylcarnitines. An accurate quantitation was achieved by the use of stable isotope-labeled internal standards (C2-C18) and acylcarnitines could be analyzed in the sub-nanomolar range. Control values for C2-C18 acylcarnitines in plasma were established. Concentrations ranged from 0.02 micromol/L for C14-acylcarnitine to 4.90 micromol/L for C2-acylcarnitine. The diagnostic suitability of the method was demonstrated for patients with medium-chain acyl-CoA dehydrogenase deficiency and very long-chain acyl-CoA dehydrogenase deficiency.
Sigma-Aldrich
Myristoyl-L-carnitine, ≥97.0% (HPLC)
Sigma-Aldrich
Octanoyl-L-carnitine, ≥97.0% (TLC)
Sigma-Aldrich
Decanoyl-L-carnitine, ≥94.0% (HPLC)
Sigma-Aldrich
Isovaleryl-L-carnitine, ≥94.0% (HPLC)
Sigma-Aldrich
Lauroyl-L-carnitine, ≥95.0% (HPLC)
Sigma-Aldrich
Butyryl-L-carnitine, ≥97.0% (TLC)
Sigma-Aldrich
Isobutyryl-L-carnitine, ≥97.0% (HPLC)
Sigma-Aldrich
Palmitoyl-L-carnitine, ≥97.0% (HPLC)
Sigma-Aldrich
Stearoyl-L-carnitine, ≥97.0% (TLC)