MilliporeSigma
  • gp130 and c-Kit signalings synergize for ex vivo expansion of human primitive hemopoietic progenitor cells.
gp130 and c-Kit signalings synergize for ex vivo expansion of human primitive hemopoietic progenitor cells.
Proceedings of the National Academy of Sciences of the United States of America (1995-03-28)
X Sui, K Tsuji, R Tanaka, S Tajima, K Muraoka, Y Ebihara, K Ikebuchi, K Yasukawa, T Taga, T Kishimoto
ABSTRACT
gp130, a signal-transducing receptor component of interleukin 6 (IL-6), associates with an IL-6 and IL-6 receptor (IL-6) complex and transduces signals. To examine the role of gp130 signaling in the expansion of human hemopoietic progenitor cells, we tested the effects of a recombinant soluble human IL-6 receptor (sIL-6R) and/or IL-6 in combination with other cytokines on purified human umbilical cord blood CD34+ cells, using methylcellulose clonal assay and suspension culture in the presence or absence of serum. A combination of sIL-6R and IL-6 (sIL-6R/IL-6), but not sIL-6R or IL-6 alone, was found to dramatically stimulate expansion of hemopoietic progenitor cells as well as CD34+ cells in the presence of stem cell factor. Significant generation of multipotential hemopoietic progenitors over a period of 3 weeks in suspension culture and efficient formation of colonies, especially multilineage and blast cell colonies, in methylcellulose assay supplemented with a combination of sIL-6R/IL-6 together with stem cell factor were observed in serum-containing and serum-free culture. Addition of anti-gp130 monoclonal antibodies or anti-IL-6R monoclonal antibodies to the above cultures dose-dependently inhibited the expansion of progenitor cells in suspension culture and also completely blocked the formation of multilineage colonies in methylcellulose culture. These findings demonstrated that the significant expansion of human primitive hemopoietic progenitors could be achieved with the gp130 and c-Kit signalings initiated by the sIL-6R/IL-6 complex in the presence of stem cell factor and suggested the possible application of this method for ex vivo expansion of CD34+ cells for bone marrow transplantation.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)