MilliporeSigma
  • Oligosaccharide sensing with chromophore-modified curdlan in aqueous media.
Oligosaccharide sensing with chromophore-modified curdlan in aqueous media.
Chemical communications (Cambridge, England) (2010-11-10)
Gaku Fukuhara, Yoshihisa Inoue
ABSTRACT
A newly synthesized chromophore-modified curdlan functions as a saccharide chemosensor in aqueous solution, enabling us to discriminate tetrasaccharide acarbose from 24 mono-, di-, tri-, and tetrasaccharides.
Product Description
Supelco
4-(Dimethylamino)benzoyl chloride, LiChropur, ≥99.0% (HPLC)