MilliporeSigma
  • Equilibrium dialysis, ultrafiltration, and ultracentrifugation compared for determining the plasma-protein-binding characteristics of valproic acid.
Equilibrium dialysis, ultrafiltration, and ultracentrifugation compared for determining the plasma-protein-binding characteristics of valproic acid.
Clinical chemistry (1985-01-01)
J Barré, J M Chamouard, G Houin, J P Tillement
ABSTRACT
Equilibrium dialysis, ultrafiltration, and ultracentrifugation were compared to determine their reliability and applicability in the study of binding of an anticonvulsant drug, valproic acid, by plasma proteins. We studied drug binding with pooled serum and with solutions of human serum albumin at physiological concentrations. We compared binding characteristics such as number of binding sites, affinity constants, and percent of binding as measured by each method in the therapeutic range for valproic acid. Results by ultracentrifugation differed from those by equilibrium dialysis and ultrafiltration, which agreed reasonably well with each other.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially fatty acid free