MilliporeSigma
  • The adaptor ASC has extracellular and 'prionoid' activities that propagate inflammation.
The adaptor ASC has extracellular and 'prionoid' activities that propagate inflammation.
Nature immunology (2014-06-24)
Bernardo S Franklin, Lukas Bossaller, Dominic De Nardo, Jacqueline M Ratter, Andrea Stutz, Gudrun Engels, Christoph Brenker, Mark Nordhoff, Sandra R Mirandola, Ashraf Al-Amoudi, Matthew S Mangan, Sebastian Zimmer, Brian G Monks, Martin Fricke, Reinhold E Schmidt, Terje Espevik, Bernadette Jones, Andrew G Jarnicki, Philip M Hansbro, Patricia Busto, Ann Marshak-Rothstein, Simone Hornemann, Adriano Aguzzi, Wolfgang Kastenmüller, Eicke Latz
ABSTRACT
Microbes or danger signals trigger inflammasome sensors, which induce polymerization of the adaptor ASC and the assembly of ASC specks. ASC specks recruit and activate caspase-1, which induces maturation of the cytokine interleukin 1β (IL-1β) and pyroptotic cell death. Here we found that after pyroptosis, ASC specks accumulated in the extracellular space, where they promoted further maturation of IL-1β. In addition, phagocytosis of ASC specks by macrophages induced lysosomal damage and nucleation of soluble ASC, as well as activation of IL-1β in recipient cells. ASC specks appeared in bodily fluids from inflamed tissues, and autoantibodies to ASC specks developed in patients and mice with autoimmune pathologies. Together these findings reveal extracellular functions of ASC specks and a previously unknown form of cell-to-cell communication.
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Anti-ASC Antibody, clone 2EI-7, clone 2EI-7, from mouse
Sigma-Aldrich
Triton X-100, laboratory grade