MilliporeSigma
  • Induction of pluripotent stem cells from human third molar mesenchymal stromal cells.
Induction of pluripotent stem cells from human third molar mesenchymal stromal cells.
The Journal of biological chemistry (2010-07-03)
Yasuaki Oda, Yasuhide Yoshimura, Hiroe Ohnishi, Mika Tadokoro, Yoshihiro Katsube, Mari Sasao, Yoko Kubo, Koji Hattori, Shigeru Saito, Katsuhisa Horimoto, Shunsuke Yuba, Hajime Ohgushi
ABSTRACT
The expression of four transcription factors (OCT3/4, SOX2, KLF4, and MYC) can reprogram mouse as well as human somatic cells to induced pluripotent stem (iPS) cells. We generated iPS cells from mesenchymal stromal cells (MSCs) derived from human third molars (wisdom teeth) by retroviral transduction of OCT3/4, SOX2, and KLF4 without MYC, which is considered as oncogene. Interestingly, some of the clonally expanded MSCs could be used for iPS cell generation with 30-100-fold higher efficiency when compared with that of other clonally expanded MSCs and human dermal fibroblasts. Global gene expression profiles demonstrated some up-regulated genes regarding DNA repair/histone conformational change in the efficient clones, suggesting that the processes of chromatin remodeling have important roles in the cascade of iPS cells generation. The generated iPS cells resembled human embryonic stem (ES) cells in many aspects, including morphology, ES marker expression, global gene expression, epigenetic states, and the ability to differentiate into the three germ layers in vitro and in vivo. Because human third molars are discarded as clinical waste, our data indicate that clonally expanded MSCs derived from human third molars are a valuable cell source for the generation of iPS cells.
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Leukocyte Alkaline Phosphatase Kit, based on naphthol AS-BI and fast red violet LB