MilliporeSigma
  • Isolation of a human laminin B2 (LAMB2) cDNA clone and assignment of the gene to chromosome region 1q25----q31.
Isolation of a human laminin B2 (LAMB2) cDNA clone and assignment of the gene to chromosome region 1q25----q31.
Cytogenetics and cell genetics (1988-01-01)
Y Fukushima, T Pikkarainen, T Kallunki, R L Eddy, M G Byers, L L Haley, W M Henry, K Tryggvason, T B Shows
ABSTRACT
Laminin B2 is one of the three polypeptide chains of laminin, a large, complex glycoprotein synthesized by a variety of cells and specifically deposited in basement membranes. A cloned cDNA that encodes the human laminin B2 chain was isolated and characterized. The human laminin B2 gene (LAMB2) was assigned to region 1q25----q31 by (1) hybridization of the probe to DNA from a panel of human x mouse somatic cell hybrids containing different human chromosomes and (2) in situ hybridization to isolated metaphase chromosomes.