MilliporeSigma
  • A comprehensive mechanism of fibrin network formation involving early branching and delayed single- to double-strand transition from coupled time-resolved X-ray/light-scattering detection.
A comprehensive mechanism of fibrin network formation involving early branching and delayed single- to double-strand transition from coupled time-resolved X-ray/light-scattering detection.
Journal of the American Chemical Society (2014-03-25)
Mattia Rocco, Matteo Molteni, Marco Ponassi, Guido Giachi, Marco Frediani, Alexandros Koutsioubas, Aldo Profumo, Didier Trevarin, Barbara Cardinali, Patrice Vachette, Fabio Ferri, Javier Pérez
ABSTRACT
The formation of a fibrin network following fibrinogen enzymatic activation is the central event in blood coagulation and has important biomedical and biotechnological implications. A non-covalent polymerization reaction between macromolecular monomers, it consists basically of two complementary processes: elongation/branching generates an interconnected 3D scaffold of relatively thin fibrils, and cooperative lateral aggregation thickens them more than 10-fold. We have studied the early stages up to the gel point by fast fibrinogen:enzyme mixing experiments using simultaneous small-angle X-ray scattering and wide-angle, multi-angle light scattering detection. The coupled evolutions of the average molecular weight, size, and cross section of the solutes during the fibrils growth phase were thus recovered. They reveal that extended structures, thinner than those predicted by the classic half-staggered, double-stranded mechanism, must quickly form. Following extensive modeling, an initial phase is proposed in which single-bonded "Y-ladder" polymers rapidly elongate before undergoing a delayed transition to the double-stranded fibrils. Consistent with the data, this alternative mechanism can intrinsically generate frequent, random branching points in each growing fibril. The model predicts that, as a consequence, some branches in these expanding "lumps" eventually interconnect, forming the pervasive 3D network. While still growing, other branches will then undergo a Ca(2+)/length-dependent cooperative collapse on the resulting network scaffolding filaments, explaining their sudden thickening, low final density, and basic mechanical properties.
Sigma-Aldrich
Syringe PP/PE without needle, luer slip tip, eccentric, capacity 20 mL, graduated, 1 mL, sterile
Corning® syringe filters, cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.2 μm