MilliporeSigma
  • Cloning and molecular analysis of a mannitol operon of phosphoenolpyruvate-dependent phosphotransferase (PTS) type from Vibrio cholerae O395.
Cloning and molecular analysis of a mannitol operon of phosphoenolpyruvate-dependent phosphotransferase (PTS) type from Vibrio cholerae O395.
Archives of microbiology (2010-12-25)
Sanath Kumar, Kenneth P Smith, Jody L Floyd, Manuel F Varela
ABSTRACT
A putative mannitol operon of the phosphoenolpyruvate phosphotransferase (PTS) type was cloned from Vibrio cholerae O395, and its activity was studied in Escherichia coli. The 3.9-kb operon comprising three genes is organized as mtlADR. Based on the sequence analysis, these were identified as genes encoding a putative mannitol-specific enzyme IICBA (EII(Mtl)) component (MtlA), a mannitol-1-phosphate dehydrogenase (MtlD), and a mannitol operon repressor (MtlR). The transport of [(3)H]mannitol by the cloned mannitol operon in E. coli was 13.8 ± 1.4 nmol/min/mg protein. The insertional inactivation of EII(Mtl) abolished mannitol and sorbitol transport in V. cholerae O395. Comparison of the mannitol utilization apparatus of V. cholerae with those of Gram-negative and Gram-positive bacteria suggests highly conserved nature of the system. MtlA and MtlD exhibit 75% similarity with corresponding sequences of E. coli mannitol operon genes, while MtlR has 63% similarity with MtlR of E. coli. The cloning of V. cholerae mannitol utilization system in an E. coli background will help in elucidating the functional properties of this operon.