MilliporeSigma
  • Mono- and binuclear copper(II) complexes of new hydrazone ligands derived from 4,6-diacetylresorcinol: Synthesis, spectral studies and antimicrobial activity.
Mono- and binuclear copper(II) complexes of new hydrazone ligands derived from 4,6-diacetylresorcinol: Synthesis, spectral studies and antimicrobial activity.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2014-03-13)
Magdy Shebl, Mosad A El-ghamry, Saied M E Khalil, Mona A A Kishk
ABSTRACT
Two new hydrazone ligands, H2L(1) and H2L(2), were synthesized by the condensation of 4,6-diacetylresorcinol with 3-hydrazino-5,6-diphenyl-1,2,4-triazine and isatin monohydrazone, respectively. The structures of the ligands were elucidated by elemental analyses, IR, (1)H NMR, electronic and mass spectra. Reactions of the ligands with several copper(II) salts, including AcO(-), NO3(-), SO4(2-), Cl(-) and Br(-) afforded mono- and binuclear metal complexes. Also, the ligands were allowed to react with Cu(II) ion in the presence of a secondary ligand (L') [N,O-donor; 8-hydroxyquinoline, N,N-donor; 1,10-phenanthroline or O,O-donor; benzoylacetone]. Characterization and structure elucidation of the prepared complexes were achieved by elemental and thermal analyses, IR, electronic, mass and ESR spectra as well as conductivity and magnetic susceptibility measurements. The ESR spin Hamiltonian parameters of some complexes were calculated. The spectroscopic data showed that the H2L(1) ligand acts as a neutral or monobasic tridentate ligand while the H2L(2) ligand acts as a bis(monobasic tridentate) ligand. The coordination sites with the copper(II) ion are phenolic oxygen, azomethine nitrogen and triazinic nitrogen (H2L(1) ligand) or isatinic oxygen (H2L(2) ligand). The metal complexes exhibited octahedral and square planar geometrical arrangements depending on the nature of the anion. The ligands and some metal complexes showed antimicrobial activity.
Sigma-Aldrich
1,1′-(4,6-Dihydroxy-1,3-phenylene)bisethanone, 99%