MilliporeSigma
  • Characterization of multiple mRNAs that encode mammalian translation initiation factor 5 (eIF-5).
Characterization of multiple mRNAs that encode mammalian translation initiation factor 5 (eIF-5).
The Journal of biological chemistry (1996-07-12)
K Si, K Das, U Maitra
ABSTRACT
Eukaryotic translation initiation factor 5 (eIF-5) interacts with the 40 S initiation complex (40S.mRNA.MettRNAf.eIF-2.GTP) to promote the hydrolysis of bound GTP with the concomitant joining of the 60 S ribosomal subunit to the 40 S initiation complex to form a functional 80 S initiation complex. In this paper, the multiple mRNAs that encode mammalian eIF-5 have been characterized. In rat tissues, three major eIF-5 mRNAs of 3.5, 2.8, and 2.2 kilobases in length are detected. All major eIF-5 mRNAs are initiated from a single transcription initiation site, contain identical 5'-untranslated and coding regions, but differ from one another only in the length of their 3'-untranslated regions. The different lengths of the 3'-untranslated region of eIF-5 mRNAs are generated by the use of alternative polyadenylation signals. Additionally, we demonstrate tissue-specific variations in eIF-5 mRNA expression as well as preference for polyadenylation sites. These results should lead to increased understanding of the regulation of eIF-5 gene expression.