MilliporeSigma
  • Further immunohistochemical characterization of BRD1 a new susceptibility gene for schizophrenia and bipolar affective disorder.
Further immunohistochemical characterization of BRD1 a new susceptibility gene for schizophrenia and bipolar affective disorder.
Brain structure & function (2009-09-19)
Carsten Reidies Bjarkam, Thomas J Corydon, Inger Marie L Olsen, Jonatan Pallesen, Mette Nyegaard, Tue Fryland, Ole Mors, Anders D Børglum
ABSTRACT
We have recently shown that the gene BRD1 is associated with schizophrenia and bipolar affective disorder and that the BRD1 protein (BRD1) which is expressed in neurons may occur in a short and a long variant. The aim of the study was to generate polyclonal antibodies against new BRD1 epitopes enabling discrimination between the long and short BRD1 variants, and elucidate the BRD1 distribution in several human tissues, including the CNS. Polyclonal rabbit antibodies were raised against three different BRD1 epitopes. One (67) was specific for the long BRD1 variant, whereas the two others (63/64 and 65/66) like the original monoclonal mouse antibody (K22) were predicted to stain both variants. Immunohistochemical staining procedures were subsequently performed on paraffin-embedded human cerebral cortex and microarray slides containing 30 different human tissues. Western blotting confirmed the predicted specificity of the developed antibodies. K22, 63/64 and 65/66 displayed a similar neuronal staining pattern characterized by a distinct but weak nuclear staining, while the surrounding cytoplasm and proximal dendrites were more intensely stained. Interestingly, staining with 67 generated in contrast primarily an intense nuclear staining. The new antibodies resulted, furthermore, in a prominent neuroglial reaction characterized by staining of cell bodies, nuclei and glial processes. The tissue microarray analysis revealed that BRD1 was widely distributed in human tissues. The particular expression profile, e.g., the degree of nuclear and/or cytoplasmatic staining, seemed, however, to be highly tissue dependent. These results suggest a general role of BRD1 in the cell and stress that the two BRD1 variants may play different roles in the etiology of psychiatric disease.