MilliporeSigma
  • Pleckstrin 2, a widely expressed paralog of pleckstrin involved in actin rearrangement.
Pleckstrin 2, a widely expressed paralog of pleckstrin involved in actin rearrangement.
The Journal of biological chemistry (1999-07-27)
M H Hu, E M Bauman, R L Roll, N Yeilding, C S Abrams
ABSTRACT
We have identified a cDNA for pleckstrin 2 that is 39% identical and 65% homologous to the original pleckstrin. Like the original pleckstrin 1, this protein contains a pleckstrin homology (PH) domain at each end of the molecule as well as a DEP (Dishevelled, Egl-10, and pleckstrin) domain in the intervening sequence. A Northern blot probed with the full-length cDNA reveals that this homolog is ubiquitously expressed and is most abundant in the thymus, large bowel, small bowel, stomach, and prostate. Unlike pleckstrin 1, this newly discovered protein does not contain obvious sites of PKC phosphorylation, and in transfected Cos-7 cells, it is a poor substrate for phosphorylation, even after PMA stimulation. Cells expressing pleckstrin 2 undergo a dramatic shape change associated with actin rearrangement, including a loss of central F-actin and a redistribution of actin toward the cell cortex. Overexpression of pleckstrin 2 causes large lamellipodia and peripheral ruffle formation. A variant of pleckstrin 2 lacking both PH domains still had some membrane binding but did not efficiently induce lamellipodia, suggesting that the PH domains of pleckstrin 2 contribute to lamellipodia formation. This work describes a novel, widely expressed, membrane-associating protein and suggests that pleckstrin 2 may help orchestrate cytoskeletal arrangement.