MilliporeSigma
  • Synthesis of all-conjugated diblock copolymers by quasi-living polymerization and observation of their microphase separation.
Synthesis of all-conjugated diblock copolymers by quasi-living polymerization and observation of their microphase separation.
Journal of the American Chemical Society (2008-05-30)
Yue Zhang, Keisuke Tajima, Kouske Hirota, Kazuhito Hashimoto
ABSTRACT
We designed and synthesized the all-conjugated diblock copolymers poly(3-hexylthiophene-block-3-(2-ethylhexyl)thiophene)s (P(3HT-b-3EHT)s) via a modified Grignard metathesis (GRIM), a type of quasi-living polymerization, and studied their microphase-separated structures. The P(3HT-b-3EHT)s synthesized had well-controlled molecular weights and very narrow polydispersity indices (PDIs), which demonstrates the usefulness of GRIM polymerization for the synthesis of semiconducting block copolymers. P(3HT-b-3EHT)s self-organized to form clear microphase-separated patterns upon thermal treatment, as observed by AFM. Interestingly, the enhancement of the interchain interaction of the P3HT segments compared with the P3HT homopolymer was clearly observed from the UV-vis spectra, despite the fact that the amount of crystalline P3HT fraction was reduced to 83% of the total polymer amount in P(3HT-b-3EHT). It is suggested that the relatively unconstrained, amorphous segments of P3EHT can enhance the crystallization of P3HT segments to form an ordered self-organized nanostructure.