MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Association study of matrix metalloproteinase-12 gene polymorphisms and asthma in a Japanese population.
Association study of matrix metalloproteinase-12 gene polymorphisms and asthma in a Japanese population.
International archives of allergy and immunology (2012-10-19)
Fumiya Yamaide, Siizkhuu Undarmaa, Yoichi Mashimo, Naoki Shimojo, Takayasu Arima, Yoshinori Morita, Tomomitsu Hirota, Kimie Fujita, Akihiko Miyatake, Satoru Doi, Kazuki Sato, Shuichi Suzuki, Toshiyuki Nishimuta, Hiroko Watanabe, Akira Hoshioka, Minako Tomiita, Akiko Yamaide, Misa Watanabe, Yoshitaka Okamoto, Yoichi Kohno, Mayumi Tamari, Akira Hata, Yoichi Suzuki
ABSTRACT
Matrix metalloproteinase 12 gene (MMP12) has been shown to be associated with asthma in a Caucasian population. In this study, we investigate whether single-nucleotide polymorphisms (SNPs) of MMP12 are associated with a risk for asthma in a Japanese population. We tested for an association between SNPs in MMP12 and asthma, including its severity, in a Japanese population (630 pediatric and 417 adult patients with atopic asthma and 336 children and 632 adults as controls). The rs652438 A and G variants (N357S) were generated by site-directed mutagenesis and an assay with artificial peptide substrates was used to compare two types of MMP12 activity. The effect of MMP12 inhibition with MMP12-specific small interfering RNA (siRNA) on chemokine secretion from airway epithelial cells was also tested in vitro. N357S showed a p value <0.05 for childhood and combined (adult plus childhood) asthma in the dominant model [odds ratio (OR) 1.60, 95% confidence interval (CI) 1.00-2.56, p = 0.047; OR 1.40, 95% CI 1.04-1.89, p = 0.028, respectively]. This risk variant is associated with asthma severity in adult patients. In the functional assay, the minor-allele enzyme showed significantly lower activity than the major-allele enzyme. MMP12-specific siRNA suppressed IP-10 secretion from airway epithelial cells upon stimulation with IFN-β. Our results suggest that MMP12 confers susceptibility to asthma and is associated with asthma severity in a Japanese population. MMP12 may be associated with asthma through inappropriate attraction of leukocytes to the inflamed tissue.