MilliporeSigma
  • Exclusive selectivity in the one-pot formation of C-C and C-Se bonds involving Ni-catalyzed alkyne hydroselenation: optimization of the synthetic procedure and a mechanistic study.
Exclusive selectivity in the one-pot formation of C-C and C-Se bonds involving Ni-catalyzed alkyne hydroselenation: optimization of the synthetic procedure and a mechanistic study.
The Journal of organic chemistry (2014-10-08)
Nikolay V Orlov, Igor V Chistyakov, Levon L Khemchyan, Valentine P Ananikov, Irina P Beletskaya, Zoya A Starikova
ABSTRACT
A unique Ni-catalyzed transformation is reported for the one-pot highly selective synthesis of previously unknown monoseleno-substituted 1,3-dienes starting from easily available terminal alkynes and benzeneselenol. The combination of a readily available catalyst precursor, Ni(acac)2, and an appropriately tuned phosphine ligand, PPh2Cy, resulted in the exclusive assembly of the s-gauche diene skeleton via the selective formation of C-C and C-Se bonds. The unusual diene products were stable under regular experimental conditions, and the products maintained the s-gauche geometry both in the solid state and in solution, as confirmed by X-ray analysis and NMR spectroscopy. Thorough mechanistic studies using ESI-MS revealed the key Ni-containing species involved in the reaction.