MilliporeSigma
  • In vitro effect of multiple hydrogen peroxide gas plasma sterilizations on the rate of closure of ameroid constrictors.
In vitro effect of multiple hydrogen peroxide gas plasma sterilizations on the rate of closure of ameroid constrictors.
American journal of veterinary research (2014-09-26)
Elizabeth M Thompson, Heather A Towle Millard, George E Moore, Lynn Guptill
ABSTRACT
To evaluate the effect of multiple hydrogen peroxide gas plasma (HPGP) sterilizations on the rate of closure of ameroid constrictors. Thirty-six 5.0-mm ameroid constrictors. Ameroid constrictors were randomly allocated to 6 groups. Each group underwent 1, 2, 3, 4, 5, or 6 HPGP sterilizations. Ameroid constrictors were then incubated for 35 days in canine plasma and digitally imaged at predetermined times during incubation. One individual, who was unaware of the group to which each ameroid constrictor was assigned, measured the lumen area of the constrictor on each digital image. Mean lumen area was compared among groups. No ameroid constrictors were completely closed after 35 days of incubation in canine plasma. Mean lumen area after incubation did not differ among constrictors that underwent 1, 2, and 3 sterilizations. Constrictors that underwent 4 sterilizations were closed significantly more than were those that underwent 1, 2, or 3 sterilizations. Mean lumen area after incubation did not differ significantly between constrictors that underwent 5 and 6 sterilizations, although the final lumen areas for those constrictors were significantly smaller than those for constrictors that underwent 1, 2, 3, and 4 sterilizations. Ameroid constrictors that underwent 5 and 6 HPGP sterilizations had a 9% to 12% decrease in lumen area, compared with that of constrictors that underwent ≤ 4 plasma sterilizations, and the use of such constrictors could increase the risk of portal hypertension and secondary acquired shunting or decrease the risk of persistent shunting.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)