MilliporeSigma
  • Conversion of 2-furylcarbinols with alkyl or aryl azides to highly functionalized 1,2,3-triazoles via cascade formal [3 + 2] cycloaddition/ring-opening.
Conversion of 2-furylcarbinols with alkyl or aryl azides to highly functionalized 1,2,3-triazoles via cascade formal [3 + 2] cycloaddition/ring-opening.
Organic letters (2014-09-16)
Jiawei Guo, Binxun Yu, Ya-Nan Wang, Dongyu Duan, Li-Li Ren, Ziwei Gao, Jing Gou
ABSTRACT
A Lewis acid promoted cascade cycloaddition/ring-opening of 2-furylcarbinols with alkyl or aryl azides is described. The reaction features an initial formal [3 + 2] cycloaddition to form a trisubstitued triazole motif, followed by a ring opening of furan to generate the (E)-configuration of the enone. A wide range of highly functionalized triazoles is expediently and efficiently synthesized in a highly step-economical manner.
Sigma-Aldrich
Furfuryl alcohol, ≥97%, FG
Sigma-Aldrich
Furfuryl alcohol, 98%
Sigma-Aldrich
Furfuryl alcohol, natural, ≥95%, FG
Supelco
Furfuryl alcohol, analytical standard