MilliporeSigma
  • Patched-1 proapoptotic activity is downregulated by modification of K1413 by the E3 ubiquitin-protein ligase Itchy homolog.
Patched-1 proapoptotic activity is downregulated by modification of K1413 by the E3 ubiquitin-protein ligase Itchy homolog.
Molecular and cellular biology (2014-08-06)
Xiaole L Chen, Pilar Chinchilla, Joanna Fombonne, Lan Ho, Catherine Guix, James H Keen, Patrick Mehlen, Natalia A Riobo
ABSTRACT
The Hedgehog (Hh) receptor Patched-1 (PTCH1) opposes the activation of Gli transcription factors and induces cell death through a Gli-independent pathway. Here, we report that the C-terminal domain (CTD) of PTCH1 interacts with and is ubiquitylated on K1413 by the E3 ubiquitin-protein ligase Itchy homolog (Itch), a Nedd4 family member. Itch induces the ubiquitylation of K1413, the reduction of PTCH1 levels at the plasma membrane, and degradation, activating Gli transcriptional activity in the absence of Hh ligands. Silencing of Itch stabilizes PTCH1 and increases its level of retention at the plasma membrane. Itch is the preferential PTCH1 E3 ligase in the absence of Hh ligands, since of the other seven Nedd4 family members, only WW domain-containing protein 2 (WWP2) showed a minor redundant role. Like Itch depletion, mutation of the ubiquitylation site (K1314R) resulted in the accumulation of PTCH1 at the plasma membrane, prolongation of its half-life, and increased cell death by hyperactivation of caspase-9. Remarkably, Itch is the main determinant of PTCH1 stability under resting conditions but not in response to Sonic Hedgehog. In conclusion, our findings reveal that Itch is a key regulator of ligand-independent Gli activation and noncanonical Hh signaling by the governance of basal PTCH1 internalization and degradation.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
L-Lysine, analytical standard
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology