MilliporeSigma
  • Contaminant-specific targeting of olfactory sensory neuron classes: connecting neuron class impairment with behavioural deficits.
Contaminant-specific targeting of olfactory sensory neuron classes: connecting neuron class impairment with behavioural deficits.
Chemosphere (2014-03-19)
William A Dew, Ali Azizishirazi, Greg G Pyle
ABSTRACT
The olfactory system of fish comprises several classes of olfactory sensory neurons (OSNs). The odourants L-alanine and taurocholic acid (TCA) specifically activate microvillous or ciliated OSNs, respectively, in fish. We recorded electro-olfactograms (EOG) in fathead minnows (Pimephales promelas; a laboratory-reared model species) and wild yellow perch (Perca flavescens) whose olfactory chambers were perfused with either L-alanine or TCA to determine if OSN classes were differentially vulnerable to contaminants, in this case copper or nickel. Results were consistent in both species and demonstrated that nickel targeted and impaired microvillous OSN function, while copper targeted and impaired ciliated OSN function. This result suggests that contaminant-specific effects observed in model laboratory species extrapolate to wild fish populations. Moreover, fathead minnows exposed to copper failed to perceive a conspecific alarm cue in a choice maze, whereas those exposed to nickel could respond to the same conspecific cue. These results demonstrate that fathead minnows perceive conspecific, damage-released alarm cue by ciliated, but not microvillous, OSNs. Fish living in copper-contaminated environments may be more vulnerable to predation than those in clean lakes owing to targeted effects on ciliated OSNs.
Product Description
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Nickel, foil, 25mm disks, thickness 0.025mm, annealed, 99%
Nickel, tube, 100mm, outside diameter 3.0mm, inside diameter 2.4mm, wall thickness 0.3mm, hard, 99.5%
Nickel, wire reel, 20m, diameter 1.0mm, hard, 99.98%
Nickel, tube, 200mm, outside diameter 5.9mm, inside diameter 5.65mm, wall thickness 0.125mm, hard, 99.5%
Nickel, tube, 200mm, outside diameter 0.8mm, inside diameter 0.64mm, wall thickness 0.08mm, as drawn, 99.5%
Nickel, tube, 1000mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, wire reel, 5m, diameter 0.5mm, as drawn, 99%
Nickel, tube, 1000mm, outside diameter 0.8mm, inside diameter 0.3mm, wall thickness 0.25mm, as drawn, 99.5%
Nickel, foil, 15mm disks, thickness 0.5mm, as rolled, 99.99+%
Nickel, foil, 25mm disks, thickness 0.15mm, annealed, 99%
Nickel, wire reel, 50m, diameter 0.025mm, as drawn, 99.6+%
Nickel, foil, 20m coil, thickness 0.125mm, coil width 1mm, as rolled, 99%
Nickel, tube, 100mm, outside diameter 0.75mm, inside diameter 0.59mm, wall thickness 0.08mm, hard, 99.5%
Nickel, wire reel, 20m, diameter 2.0mm, hard, 99.98%
Nickel, foil, 15mm disks, thickness 0.20mm, annealed, 99%
Nickel, foil, 1m coil, thickness 0.15mm, annealed, 99%
Nickel, wire reel, 5m, diameter 0.5mm, hard, 99.98%