MilliporeSigma
  • Molecular characterization of long-term survivors of glioblastoma using genome- and transcriptome-wide profiling.
Molecular characterization of long-term survivors of glioblastoma using genome- and transcriptome-wide profiling.
International journal of cancer (2014-03-13)
Guido Reifenberger, Ruthild G Weber, Vera Riehmer, Kerstin Kaulich, Edith Willscher, Henry Wirth, Jens Gietzelt, Bettina Hentschel, Manfred Westphal, Matthias Simon, Gabriele Schackert, Johannes Schramm, Jakob Matschke, Michael C Sabel, Dorothee Gramatzki, Jörg Felsberg, Christian Hartmann, Joachim P Steinbach, Uwe Schlegel, Wolfgang Wick, Bernhard Radlwimmer, Torsten Pietsch, Jörg C Tonn, Andreas von Deimling, Hans Binder, Michael Weller, Markus Loeffler
ABSTRACT
The prognosis of glioblastoma, the most malignant type of glioma, is still poor, with only a minority of patients showing long-term survival of more than three years after diagnosis. To elucidate the molecular aberrations in glioblastomas of long-term survivors, we performed genome- and/or transcriptome-wide molecular profiling of glioblastoma samples from 94 patients, including 28 long-term survivors with >36 months overall survival (OS), 20 short-term survivors with <12 months OS and 46 patients with intermediate OS. Integrative bioinformatic analyses were used to characterize molecular aberrations in the distinct survival groups considering established molecular markers such as isocitrate dehydrogenase 1 or 2 (IDH1/2) mutations, and O(6) -methylguanine DNA methyltransferase (MGMT) promoter methylation. Patients with long-term survival were younger and more often had IDH1/2-mutant and MGMT-methylated tumors. Gene expression profiling revealed over-representation of a distinct (proneural-like) expression signature in long-term survivors that was linked to IDH1/2 mutation. However, IDH1/2-wildtype glioblastomas from long-term survivors did not show distinct gene expression profiles and included proneural, classical and mesenchymal glioblastoma subtypes. Genomic imbalances also differed between IDH1/2-mutant and IDH1/2-wildtype tumors, but not between survival groups of IDH1/2-wildtype patients. Thus, our data support an important role for MGMT promoter methylation and IDH1/2 mutation in glioblastoma long-term survival and corroborate the association of IDH1/2 mutation with distinct genomic and transcriptional profiles. Importantly, however, IDH1/2-wildtype glioblastomas in our cohort of long-term survivors lacked distinctive DNA copy number changes and gene expression signatures, indicating that other factors might have been responsible for long survival in this particular subgroup of patients.
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type I, 0.5-3.0 unit/mg solid (plus numerous enzyme activities associated with porcine heart)
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in E. coli, lyophilized powder, ≥80 units/mg protein