MilliporeSigma
  • Characterization and tissue-specific expression of human GSK-3-binding proteins FRAT1 and FRAT2.
Characterization and tissue-specific expression of human GSK-3-binding proteins FRAT1 and FRAT2.
Gene (2002-07-04)
Sarah J Freemantle, Holly B Portland, Katherine Ewings, Florence Dmitrovsky, Keith DiPetrillo, Michael J Spinella, Ethan Dmitrovsky
ABSTRACT
We have isolated the entire coding sequence of human FRAT2 (frequently rearranged in advanced T-cell lymphomas-2). It exhibits appreciable amino acid identity to FRAT1 (77%) which was initially isolated as frequently being overexpressed in a murine leukemia virus insertion model in murine tumors. FRAT proteins are thought to play a role in Wnt signaling. They can bind to glycogen synthase kinase-3 (GSK-3) and Dishevelled, two proteins involved in Wnt signal transduction. Both hFRAT1 and hFRAT2 are intronless genes localized to the same portion of chromosome 10q24.1 and separated by only 10.7 kb. In a broad range of human tissues FRAT1 and FRAT2 are readily detected and expressed in a near identical pattern. Both species are repressed when the human embryonal carcinoma cell line, NT2/D1, is induced to differentiate with all-trans retinoic acid (RA). This treatment had no appreciable effect on FRAT levels in two other RA-sensitive cell lines that were not of germ cell tumor origin. The overlapping expression patterns suggest these two genes share a regulatory region. Both FRAT genes exhibited three species of mRNA, which varied in representation between tissues. When transiently overexpressed in COS-1 cells, the FRAT proteins were detected in the cytosol and concentrated in the nucleus. Both hFRAT1 and hFRAT2 are implicated in the selective modulation of GSK-3 activity via the Wnt signaling pathway. This study provides a foundation from which to examine the role these proteins play in Wnt-dependent and -independent processes.