MilliporeSigma
  • Plant cytokinesis is orchestrated by the sequential action of the TRAPPII and exocyst tethering complexes.
Plant cytokinesis is orchestrated by the sequential action of the TRAPPII and exocyst tethering complexes.
Developmental cell (2014-06-03)
Katarzyna Rybak, Alexander Steiner, Lukas Synek, Susan Klaeger, Ivan Kulich, Eva Facher, Gerhard Wanner, Bernhard Kuster, Viktor Zarsky, Staffan Persson, Farhah F Assaad
ABSTRACT
Plant cytokinesis is initiated in a transient membrane compartment, the cell plate, and completed by a process of maturation during which the cell plate becomes a cross wall. How the transition from juvenile to adult stages occurs is poorly understood. In this study, we monitor the Arabidopsis transport protein particle II (TRAPPII) and exocyst tethering complexes throughout cytokinesis. We show that their appearance is predominantly sequential, with brief overlap at the onset and end of cytokinesis. The TRAPPII complex is required for cell plate biogenesis, and the exocyst is required for cell plate maturation. The TRAPPII complex sorts plasma membrane proteins, including exocyst subunits, at the cell plate throughout cytokinesis. We show that the two tethering complexes physically interact and propose that their coordinated action may orchestrate not only plant but also animal cytokinesis.
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)