MilliporeSigma
Pd(II)-catalyzed C-H iodination using molecular I2 as the sole oxidant.
Journal of the American Chemical Society (2013-07-11)
Xiao-Chen Wang, Yi Hu, Samuel Bonacorsi, Yang Hong, Richard Burrell, Jin-Quan Yu
ABSTRACT
Pd-catalyzed ortho-C-H iodination directed by a weakly coordinating amide auxiliary using I2 as the sole oxidant was developed. This reaction is compatible with a wide range of heterocycles including pyridines, imidazoles, oxazoles, thiazoles, isoxazoles, and pyrazoles.
Sigma-Aldrich
2-Chloro-6-iodobenzoic acid, 95%