MilliporeSigma
  • Structure of a class C GPCR metabotropic glutamate receptor 1 bound to an allosteric modulator.
Structure of a class C GPCR metabotropic glutamate receptor 1 bound to an allosteric modulator.
Science (New York, N.Y.) (2014-03-08)
Huixian Wu, Chong Wang, Karen J Gregory, Gye Won Han, Hyekyung P Cho, Yan Xia, Colleen M Niswender, Vsevolod Katritch, Jens Meiler, Vadim Cherezov, P Jeffrey Conn, Raymond C Stevens
ABSTRACT
The excitatory neurotransmitter glutamate induces modulatory actions via the metabotropic glutamate receptors (mGlus), which are class C G protein-coupled receptors (GPCRs). We determined the structure of the human mGlu1 receptor seven-transmembrane (7TM) domain bound to a negative allosteric modulator, FITM, at a resolution of 2.8 angstroms. The modulator binding site partially overlaps with the orthosteric binding sites of class A GPCRs but is more restricted than most other GPCRs. We observed a parallel 7TM dimer mediated by cholesterols, which suggests that signaling initiated by glutamate's interaction with the extracellular domain might be mediated via 7TM interactions within the full-length receptor dimer. A combination of crystallography, structure-activity relationships, mutagenesis, and full-length dimer modeling provides insights about the allosteric modulation and activation mechanism of class C GPCRs.