MilliporeSigma
Total synthesis and stereochemical reassignment of (±)-indoxamycin B.
Angewandte Chemie (International ed. in English) (2012-02-22)
Oliver F Jeker, Erick M Carreira
ABSTRACT
Revised version: the first total synthesis of indoxamycin B leads to a stereochemical reassignment of the natural product. The synthetic route features an efficient carboannulation sequence to rapidly access the dihydroindenone system. Moreover, a series of Au(I)-catalyzed transformations served in the construction of the sterically congested core framework.
Sigma-Aldrich
Methyl 3,5-dimethylbenzoate, 98%