MilliporeSigma
  • The study of controlling pore size on electrospun carbon nanofibers for hydrogen adsorption.
The study of controlling pore size on electrospun carbon nanofibers for hydrogen adsorption.
Journal of colloid and interface science (2007-11-09)
Ji Sun Im, Soo-Jin Park, Tae Jin Kim, Young Ho Kim, Young-Seak Lee
ABSTRACT
Polyacrylonitrile (PAN)-based carbon nanofibers (CNFs) were prepared by using electrospinning method and heat treatment to get the media for hydrogen adsorption storage. Potassium hydroxide and zinc chloride activations were conducted to increase specific surface area and pore volume of CNFs. To investigate the relation between pore structure and the capacity of hydrogen adsorption, textural properties of activated CNFs were studied with micropore size distribution, specific surface area, and total pore volume by using BET (Brunauer-Emmett-Teller) surface analyzer apparatus and the capacity of hydrogen adsorption was evaluated by PCT (pressure-composition-temperature) hydrogen adsorption analyzer apparatus with volumetric method. The surface morphology of activated CNFs was observed by SEM (scanning electron microscope) images to investigate the surface change through activation. Even though specific surface area and total pore volume were important factors for increasing the capacity of hydrogen adsorption, the pore volume which has pore width (0.6-0.7 nm) was a much more effective factor than specific surface area and pore volume in PAN-based electrospun activated CNFs.
Sigma-Aldrich
Polyacrylonitrile, average Mv 150,000