MilliporeSigma
  • Assessment of polycrystalline graphites as sorbents for solid-phase microextraction of nonionic surfactants.
Assessment of polycrystalline graphites as sorbents for solid-phase microextraction of nonionic surfactants.
Journal of chromatography. A (2000-08-19)
R Aranda, P Kruus, R C Burk
ABSTRACT
Two polycrystalline graphites (pencil lead and glassy carbon) were used as sorbents for solid-phase microextraction of a nonionic alkylphenol ethoxylate surfactant (Triton X-100). Analyses were performed by reversed-phase HPLC-fluorescence detection. The presence of the benzene ring in the congeners of Triton X-100 also allowed their direct detection at lambda(ex) = 230 nm and lambda(em) = 310 nm. Variables such as time of adsorption, time of desorption and concentration of surfactant in water were evaluated. The method limit of detection was found to be 0.5 microg/l for Triton X-100, with a linear dynamic range of 0.5-150 microg/l. Results were compared to those obtained using polymeric fibers such as PDMS/DVB and Carbowax/TPR. The chemical resistance and low cost of the polycrystalline graphites are advantageous over commercially available SPME fibers.
Product Description
Supelco
GC Stationary Phase, phase Triton X-100, bottle of 50 g