MilliporeSigma
  • Strong expression of the neuronal transcription factor FOXP2 is linked to an increased risk of early PSA recurrence in ERG fusion-negative cancers.
Strong expression of the neuronal transcription factor FOXP2 is linked to an increased risk of early PSA recurrence in ERG fusion-negative cancers.
Journal of clinical pathology (2013-04-06)
Laura Stumm, Lia Burkhardt, Stefan Steurer, Ronald Simon, Meike Adam, Andreas Becker, Guido Sauter, Sarah Minner, Thorsten Schlomm, Hüseyin Sirma, Uwe Michl
ABSTRACT
Transcription factors of the forkhead box P (FOXP1-4) family have been implicated in various human cancer types before. The relevance and role of neuronal transcription factor FOXP2 in prostate cancer is unknown. A tissue microarray containing samples from more than 11 000 prostate cancers from radical prostatectomy specimens with clinical follow-up data was analysed for FOXP2 expression by immunohistochemistry. FOXP2 data were also compared with pre-existing ERG fusion (by fluorescence in situ hybridisation and immunohistochemistry) and cell proliferation (Ki67 labelling index) data. There was a moderate to strong FOXP2 protein expression in basal and secretory cells of normal prostatic glands. As compared with normal cells, FOXP2 expression was lost or reduced in 25% of cancers. Strong FOXP2 expression was linked to advanced tumour stage, high Gleason score, presence of lymph node metastases and early tumour recurrence (p<0.0001; each) in ERG fusion-negative, but not in ERG fusion-positive cancers. High FOXP2 expression was linked to high Ki67 labelling index (p<0.0001) in all cancers irrespective of ERG fusion status. These data demonstrate that similar high FOXP2 protein levels as in normal prostate epithelium exert a 'paradoxical' oncogenic role in 'non fusion-type' prostate cancer. It may be speculated that interaction of FOXP2 with members of pathways that are specifically activated in 'non fusion-type' cancers may be responsible for this phenomenon.
Sigma-Aldrich
Anti-FOXP2 antibody produced in rabbit, AB1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution