MilliporeSigma
  • Single-nucleotide polymorphisms of the KCNS3 gene are significantly associated with airway hyperresponsiveness.
Single-nucleotide polymorphisms of the KCNS3 gene are significantly associated with airway hyperresponsiveness.
Human genetics (2005-02-17)
Ke Hao, Tianhua Niu, Xin Xu, Zhian Fang, Xiping Xu
ABSTRACT
Airway hyperresponsiveness (AHR) is one of the major clinical symptoms and intermediate phenotypes of asthma. A recent genome-wide search for asthma quantitative trait loci has revealed a significant linkage signal between a p-terminal region of chromosome 2 and AHR. Thus, the gene encoding the potassium voltage-gated channel delayed-rectifier protein S3 (KCNS3) in this region is considered a positional candidate for asthma. We have evaluated a total of 12 single-nucleotide polymorphisms (SNPs) of the KCNS3 gene in a validation panel of 48 lymphoblastoid cell line DNA samples of Chinese origin. Three SNPs were found to be polymorphic and were tested. Two independent sets (an initial screening set and a replication set) of cases and controls from the original linkage study sample were collected. In the initial screening set, two SNPs (rs1031771 and rs1031772) showed suggestive association and were further confirmed by the replication set. In combined single-SNP analysis, the rs1031771 G allele (odds ratio=1.42, P=0.006) and rs1031772 T allele (odds ratio=1.40, P=0.018) were associated with a significantly higher risk of AHR. Haplotype analysis also detected significant association (P=0.006). Our findings suggest that SNPs located at the 3' downstream region of KCNS3 have a significant role in the etiology of AHR.