MilliporeSigma
  • Chemical behavior of dicyanocob(III)yrinic acid heptamethyl ester and cob(I)yrinic acid heptamethyl ester in some preparative experiments.
Chemical behavior of dicyanocob(III)yrinic acid heptamethyl ester and cob(I)yrinic acid heptamethyl ester in some preparative experiments.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences (1976-02-05)
H H Inhoffen, A Gossauer, K P Heise, H Laas
PMID4841
ABSTRACT
Preparations of C10 acetate, trifluoroacetate, and chloro derivatives of dicyanocobyrinic acid heptamethyl ester ('cobester') are described. The nature and properties of intermediates in the synthesis of the 10-chloro derivative are discussed, and the electronic absorption spectra of the products are presented.